Learn More
Abnova™ Human WNK4 Partial ORF (NP_115763.2, 1144 a.a. - 1243 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00065266-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the WNK family of serine-threonine protein kinases. The kinase is part of the tight junction complex in kidney cells, and regulates the balance between NaCl reabsorption and K(+) secretion. The kinase regulates the activities of several types of ion channels, cotransporters, and exchangers involved in electrolyte flux in epithelial cells. Mutations in this gene result in pseudohypoaldosteronism type IIB
Sequence: KHLSEVETLQTLQKKEIEDLYSRLGKQPPPGIVAPAAMLSSRQRRLSKGSFPTSRRNSLQRSEPPGPGIMRRNSLSGSSTGSQEQRASKGVTFAGDVGRMSpecifica
NP_115763.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KHLSEVETLQTLQKKEIEDLYSRLGKQPPPGIVAPAAMLSSRQRRLSKGSFPTSRRNSLQRSEPPGPGIMRRNSLSGSSTGSQEQRASKGVTFAGDVGRM | |
RUO | |
WNK4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
65266 | |
WNK4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PHA2B/PRKWNK4 | |
WNK4 | |
Recombinant | |
wheat germ expression system |