missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZIK1 Full-length ORF (AAI03958.1, 1 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00284307-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: MCVPVLKDILHLADLPGQKPYLVGECTNHHQHQKHHSAKKSLKRDMDRASYVKCCLFCMSLKPFRKWEVGKDLPAMLRLLRSLVFPGGKKPGTITECGEDIRSQKSHYKSGECGKASRHKHTPVYHPRVYTGKKLYECSKCGKAFRGKYSLVQHQRVHTGERPWECNECGKFFSQTSHLNDHRRIHTGERPYECSECGKLFRQNSSLVDHQKIHTGARPYECSQCGKSFSQKATLVKHQRVHTGERPYKCGECGNSFSQSAILNQHRRIHTGAKPYECGQCGKSFSQKATLIKHQRVHTGERPYKCGDCGKSFSQSSILIQHRRIHTGARPYECGQCGKSFSQKSGLIQHQVVHTGERPYECNKCGNSFSQCSSLIHHQKCHNTSpecifica
AAI03958.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119699/MGC119700/MGC119701/ZNF762 | |
ZIK1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
284307 | |
ZIK1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCVPVLKDILHLADLPGQKPYLVGECTNHHQHQKHHSAKKSLKRDMDRASYVKCCLFCMSLKPFRKWEVGKDLPAMLRLLRSLVFPGGKKPGTITECGEDIRSQKSHYKSGECGKASRHKHTPVYHPRVYTGKKLYECSKCGKAFRGKYSLVQHQRVHTGERPWECNECGKFFSQTSHLNDHRRIHTGERPYECSECGKLFRQNSSLVDHQKIHTGARPYECSQCGKSFSQKATLVKHQRVHTGERPYKCGECGNSFSQSAILNQHRRIHTGAKPYECGQCGKSFSQKATLIKHQRVHTGERPYKCGDCGKSFSQSSILIQHRRIHTGARPYECGQCGKSFSQKSGLIQHQVVHTGERPYECNKCGNSFSQCSSLIHHQKCHNT | |
RUO | |
ZIK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |