missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZKSCAN1 Full-length ORF (AAH22378.1, 1 a.a. - 563 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007586-P02.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16162002

  • 302.00€ / 10µg


Spedizione stimata
30-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: MMTAESREATGLSPQAAQEKDGIVIAKVEEEDEEDHMWGQDSTLQDTPPPDPEIFRQRFRRFCYQNTFGPREALSRLKELCHQWLRPEINTKEQILELLVLEQFLSILPKELQVWLQEYRPDSGEEAVTLLEDLELDLSGQQVPGQVHGPEMLARGMVPLDPVQESSSFDLHHEATQSHFKHSSRKPRLLQSRALPAAHIPAPPHEGSPRDQAMASALFTADSQAMVKIEDMAVSLILEEWGCQNLARRNLSRDNRQENYGSAFPQGGENRNENEESTSKAETSEDSASRGETTGRSQKEFGEKRDQEGKTGERQQKNPEEKTRKEKRDSGPAIGKDKKTITGERGPREKGKGLGRSFSLSSNFTTPEEVPTGTKSHRCDECGKCFTRSSSLIRHKIIHTGEKPYECSECGKAFSLNSNLVLHQRIHTGEKPHECNECGKAFSHSSNLILHQRIHSGEKPYECNECGKAFSQSSDLTKHQRIHTGEKPYECSECGKAFNRNSYLILHRRIHTREKPYKCTKCGKAFTRSSTLTLHHRIHARERASEYSPASLDAFGAFLKSCV
Specifica

Specifica

AAH22378.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
90kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
9130423L19Rik|KOX18|MGC138429|PHZ-37|ZNF139|ZNF36
ZKSCAN1
Yes
wheat germ expression system
Liquid
7586
ZKSCAN1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMTAESREATGLSPQAAQEKDGIVIAKVEEEDEEDHMWGQDSTLQDTPPPDPEIFRQRFRRFCYQNTFGPREALSRLKELCHQWLRPEINTKEQILELLVLEQFLSILPKELQVWLQEYRPDSGEEAVTLLEDLELDLSGQQVPGQVHGPEMLARGMVPLDPVQESSSFDLHHEATQSHFKHSSRKPRLLQSRALPAAHIPAPPHEGSPRDQAMASALFTADSQAMVKIEDMAVSLILEEWGCQNLARRNLSRDNRQENYGSAFPQGGENRNENEESTSKAETSEDSASRGETTGRSQKEFGEKRDQEGKTGERQQKNPEEKTRKEKRDSGPAIGKDKKTITGERGPREKGKGLGRSFSLSSNFTTPEEVPTGTKSHRCDECGKCFTRSSSLIRHKIIHTGEKPYECSECGKAFSLNSNLVLHQRIHTGEKPHECNECGKAFSHSSNLILHQRIHSGEKPYECNECGKAFSQSSDLTKHQRIHTGEKPYECSECGKAFNRNSYLILHRRIHTREKPYKCTKCGKAFTRSSTLTLHHRIHARERASEYSPASLDAFGAFLKSCV
RUO
ZKSCAN1
Wheat Germ (in vitro)
GST
Videos
SDS
Documenti