missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF230 Full-length ORF (NP_006291.2, 1 a.a. - 474 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007773-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16162112

  • 458.00€ / 25µg


Spedizione stimata
19-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: MTTFKEAVTFKDVAVFFTEEELGLLDPAQRKLYQDVMLENFTNLLSVGHQPFHPFHFLREEKFWMMETATQREGNSGGKTIAEAGPHEDCPCQQIWEQTASDLTQSQDSIINNSHFFEQGDVPSQVEAGLSIIHTGQKPSQNGKCKQSFSDVAIFDPPQQFHSGEKSHTCNECGKSFCYISALRIHQRVHLREKLSKCDMRGKEFSQSSCLQTRERVHTGEKPFKCEQCGKGFRCRAILQVHCKLHTGEKPYICEKCGRAFIHDFQLQKHQIIHTGEKPFKCEICGKSFCLRSSLNRHCMVHTAEKLYKSEECGKGFTDSLDLHKHQIIHTGQKPYNCKECGKSFRWSSYLLIHQRIHSGEKPYRCEECGKGYISKSGLNLHQRVHTGERPYNCKECGKSFSRASSILNHKKLHCRKKPFKCEDCGKRLVHRSFCKDQQGDHNGENSSKCEDCGKRYKRRLNLDIILSLFLNDM
Specifica

Specifica

NP_006291.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FDZF2
ZNF230
Yes
wheat germ expression system
Liquid
7773
ZNF230 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTTFKEAVTFKDVAVFFTEEELGLLDPAQRKLYQDVMLENFTNLLSVGHQPFHPFHFLREEKFWMMETATQREGNSGGKTIAEAGPHEDCPCQQIWEQTASDLTQSQDSIINNSHFFEQGDVPSQVEAGLSIIHTGQKPSQNGKCKQSFSDVAIFDPPQQFHSGEKSHTCNECGKSFCYISALRIHQRVHLREKLSKCDMRGKEFSQSSCLQTRERVHTGEKPFKCEQCGKGFRCRAILQVHCKLHTGEKPYICEKCGRAFIHDFQLQKHQIIHTGEKPFKCEICGKSFCLRSSLNRHCMVHTAEKLYKSEECGKGFTDSLDLHKHQIIHTGQKPYNCKECGKSFRWSSYLLIHQRIHSGEKPYRCEECGKGYISKSGLNLHQRVHTGERPYNCKECGKSFSRASSILNHKKLHCRKKPFKCEDCGKRLVHRSFCKDQQGDHNGENSSKCEDCGKRYKRRLNLDIILSLFLNDM
RUO
ZNF230
Wheat Germ (in vitro)
GST
Videos
Documenti