missing translation for 'onlineSavingsMsg'
Learn More

HYAL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18139-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898184

  • 121.38€ / 20µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This polyclonal antibody is raised against 29-460 aa of human HYAL4. The apparent molecular weight is larger than the calculated molecular weight of 54 kDa, which is likely due to posttranslational modification, presumably glycosylation.

Endo-hyaluronidase that degrades hyaluro to smaller oligosaccharide fragments. Has also chondroitin sulfate hydrolase activity, The best substrate being the galactosaminidic linkage in the sequence of a trisulfated tetrasaccharide.
Specifica

Specifica

HYAL4
Polyclonal
Unconjugated
HYAL4
Hyal 4, HYAL4, Hyaluronidase 4, hyaluronoglucosaminidase 4
Rabbit
Antigen Affinity Chromatography
RUO
23553, 404783, 77042
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q05A56, Q2M3T9
Hyal4
HYAL4 Fusion Protein Ag12766
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali