Learn More
Cytiva HyClone™ Ham's Nutrient Mixture F12 Media
Codice prodotto. 11968075
Cambia vista
Click to view available options
Quantità:
1000 mL
500 mL
6 x 500 mL
Con additivi:
Glutamine
Glutamine, Phenol Red, Sodium Pyruvate
Dimensione della confezione:
1000mL
500mL
pezzi 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Con additivi unitSize
11968075 6 x 500 mL Glutamine pezzi 6
10235122 1000 mL Glutamine 1000mL
10580273 500 mL Glutamine, Phenol Red, Sodium Pyruvate 500mL
10700445 500 mL Glutamine 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 11968075 Fornitore Cytiva N. del fornitore SH30026.FS
Please to purchase this item. Need a web account? Register with us today!
Support cell culture growth with HyClone™ Ham's Nutrient Mixture F12 Media, manufactured based on compliance with current Good Manufacturing Practices (cGMP) for Medical Devices and ISO 9001 guidelines.
  • System tested with Cytiva HyClone sera to ensure product efficacy and homogeneity
  • All products are hydrated using purified process water and undergo sterile filtration
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
TRUSTED_SUSTAINABILITY
Specifica
Forma Liquid
Product Type Ham's Nutrient Mixture F12, with L-glutamine
Sterilità Sterile-filtered
Sterilization Method 0.1 μm Sterile-filtered
Con additivi Glutamine
Quantità 6 x 500 mL
Certificazione/Conformità cGMP, ISO 9001:2000
Content And Storage 2° to 8°C, away from light
Descrizione Ham's F12, 1 X, with 1.00mM L-Glutamine