Learn More
Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, EMEA Tested
Codice prodotto. 11987328
Cambia vista
Click to view available options
Elaborazione:
Irradiated
Non-Irradiated
Dimensione della confezione:
1L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Elaborazione unitSize
11987328 Irradiated 1L
16615775 Non-Irradiated 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11987328 Fornitore Cytiva N. del fornitore SH30534.04
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)
  • Contains three to four times as much available iron and transferrin as FBS or equine serum
TRUSTED_SUSTAINABILITY
Specifica
Age 16 to 22 weeks
Product Type Calf Bovine Serum
Sterilization Method Sequential-filtered, 40 nm
Elaborazione Irradiated
Concentrazione di endotossine ≤ 10 EU/mL
Concentrazione emoglobina ≤ 10 mg/dL
Integrazione ferro Yes
Linea di prodotti HyClone
Metodi di test European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests