Learn More
Carver™ Hydraulic Fluid
Product Code. 11967137
Change view
Click to view available options
Product Type:
Hydraulic Fluid
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11967137 Hydraulic Fluid 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Special packaging extra freight cost. Please contact us for quotation
Product Code. 11967137 Supplier Carver™ Supplier No. 2170
  • ONLINE EXCLUSIVE PRICE
    72.69€
    19.14€ / 500mL
    Risparmia 53.55€
    73% Sconto
 Restricted Item
Spedizione stimata: 03-05-2027
Special packaging extra freight cost. Please contact us for quotation
Specifications
Product Type Hydraulic Fluid
Product Identifier
  • MOBIL DTE 25
Supplemental information
  • MIXTURE LIST-Contain: ZINC DITHIOPHOSPHATE; CALCIUM BIS(DI C8-C10, BRANCHED, C9 RICH, ALKYLNAPTHALENESULPHONATE) ; 2,6-DI