Learn More
Emma Safety Footwear Hydro-Tec™ Comfort Inlay Sole
Product Code. 10757145
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10757145 41 1 pair
13224554 42 Each
10154214 43 1 pair
10236084 46 1 pair
10011954 44 1 pair
10747145 39 1 pair
10001964 37 1 pair
13284544 38 Each
10327674 40 1 pair
10387574 45 1 pair
10576364 48 1 pair
10433424 36 1 pair
10246084 47 1 pair
10525024 35 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10757145 Supplier Emma Safety Footwear Supplier No. MM00107041
  • ONLINE EXCLUSIVE PRICE
    11.18€
    7.22€ / 1 paia
    Risparmia 3.96€
    35% Sconto
Spedizione stimata: 09-12-2026
Anatomically shaped and interchangeable.
  • Anti-static (ESD) and anti-bacterial
  • Breathable, regulates and absorbs moisture
  • Washable and resilient
Specifications
Color Gray
Size 41
Style Interchangeable