Learn More
Hydroxylamine Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Hydroxylamine Hydrochloride |
| CAS | 5470-11-1 |
| Quantità | 5 g |
| Formula molecolare | ClH4NO |
| Sinonimo | Hydroxylamine, hydrochloride (1:1) (ACI), Hydroxylamine, hydrochloride (8CI, 9CI), Hydroxyamine hydrochloride, Hydroxyammonium chloride, Hydroxylamine chloride, Hydroxylamine chlorohydrate, Hydroxylammonium chloride, Oxammonium hydrochloride, Hydroxylamine hydrochloride |
| InChI Formula | InChI=1S/ClH.H3NO/c;1-2/h1H;2H,1H2 |
| SMILES | Cl.NO |
| IUPAC Name | hydroxylamine;hydrochloride |
| Molecular Weight (g/mol) | 69.49 |
| Peso formulazione | 68.9981 |