Learn More
Hydroxyzine Pamoate, TRC
Specifica
Specifica
| Materiale o nome chimico | Hydroxyzine Pamoate |
| CAS | 10246-75-0 |
| Formula molecolare | C23 H16 O6 . C21 H27 Cl N2 O2 |
| Quantità | 250 mg |
| Sinonimo | 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]ethanol (1:1) (9CI, ACI), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 2-[2-[4-(p-chloro-α-phenylbenzyl)-1-piperazinyl]ethoxy]ethanol (1:1) (8CI), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, hydroxyzine salt (6CI), Ethanol, 2-[2-[4-(p-chloro-α-phenylbenzyl)-1-piperazinyl]ethoxy]-, 4,4'-methylenebis[3-hydroxy-2-naphthoate] (1:1) (salt) (8CI), Ethanol, 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (9CI), 1-(p-Chloro-α-phenylbenzyl)-4-(2-hydroxyethoxyethyl)piperazine, pamoate, 2-(2-(4-((4-chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)ethanol 4,4'-methylenebis(3-hydroxy-2-naphthoate), Atarax P, Equipose, Hydroxyzine pamoate, Hydroxyzyne pamoate, Masmoran, Paxisitil, Paxistil, Vistaril, Vistaril pamoate |
| InChI Formula | InChI=1S/C23H16O6.C21H27ClN2O2/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;22-20-8-6-19(7-9-20)21(18-4-2-1-3-5-18)24-12-10-23(11-13-24)14-16-26-17-15-25/h1-10,24-25H,11H2,(H,26,27)(H,28,29);1-9,21,25H,10-17H2 |
| SMILES | OCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3.OC(=O)c4cc5ccccc5c(Cc6c(O)c(cc7ccccc67)C(=O)O)c4O |
| IUPAC Name | 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]ethanol |
| Molecular Weight (g/mol) | 763.27 |
| Peso formulazione | 762.2708 |