Learn More
Safety First Aid HypaClean Alcohol Hand Sanitiser Gel with Pump Dispenser
Codice prodotto. 13177094
Cambia vista
Click to view available options
Colore:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Colore
13177094 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 13177094 Fornitore Safety First Aid N. del fornitore M6852PM
Spedizione stimata: 24-11-2026
The HypaClean Alcohol Hand Gel is an alcohol-based hand sanitising gel.
  • The hand Gel is an alcohol-based hand sanitiser effective at destroying 99.99% of most harmful viruses and bacteria instantly with its 65% (W/W) denatured ethanol solution.
  • This hand gel also includes moisturisers and Vitamin E.
  • The 500 mL pump-dispense bottle is ideal for workplace use.
  • It's a convenient way of keeping clean, safe and protected even when you don't have immediate access to soap and water.
  • This hand gel reduces the risk of disease and helps to prevent sickness by preventing the transfer of harmful viruses and bacteria.
Specifica
Product Type Hand Sanitizer
Colore Clear
Forma Gel
Quantità 500 mL