Learn More
Thermo Scientific™ HyperSep™ Retain PEP Cartridges
Codice prodotto. 15974417 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Peso del letto:
30 mg
60 mg
100 mg
150 mg
200 mg
300 mg
500 mg
600 mg
1000 mg
2000 mg
Capacità colonna (sistema metrico):
1 mL
3 mL
6 mL
6 ml
25 mL
Dimensione della confezione:
pezzi 100
pezzi 20
pezzi 30
pezzi 50
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Peso del letto Capacità colonna (sistema metrico) Dimensione della confezione
15974417 600 mg 6 mL pezzi 50
10595895 30 mg 1 mL pezzi 100
10259564 30 mg 3 mL pezzi 50
15984417 300 mg 3 mL pezzi 100
10505905 60 mg 3 mL pezzi 50
10448104 200 mg 3 mL pezzi 50
10351215 500 mg 3 mL pezzi 50
10618725 500 mg 6 ml pezzi 30
10015894 100 mg 6 mL pezzi 30
10269564 60 mg 6 mL pezzi 30
10279954 150 mg 6 mL pezzi 30
10565245 200 mg 6 mL pezzi 30
11504745 2000 mg 25 mL pezzi 20
11514745 1000 mg 25 mL pezzi 20
13406936 100 mg 3 mL pezzi 50
13436936 1000 mg 6 mL pezzi 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Codice prodotto. 15974417 Fornitore Thermo Scientific™ N. del fornitore 60107225
 Articolo soggetto a restrizioni
Spedizione stimata: 02-12-2026
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features:
Ideal for large samples, Hypersep Retain cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • Available in a wide range of chemistries and bed weights
Applications:
  • Drugs and metabolites in biological fluids
  • Peptides in serum, plasma or biological fluids
  • Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY
Specifica
Capacità colonna (sistema metrico) 6 mL
Descrizione 600 mg bed weight; 6 mL column volume
Peso del letto 600 mg
Da utilizzare con (applicazione) Retention of Polar and Nonpolar Analytes
Product Line HyperSep
Assorbente HyperSep Urea-Modified Polystyrene DVB
Tipo Solid Phase Extraction Cartridge
Volume (sistema metrico) 6 mL