Learn More
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 1-10μL
Codice prodotto. 10364513 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Assorbente:
BioBasic C18
BioBasic C4
BioBasic C8
HyperSep HILIC
HyperSep Hypercarb
HyperSep Hypercarb and C18 (Mix Mode)
HyperSep Poros SAX
HyperSep Poros SCX
HyperSep Poros WAX
HyperSep Titanium Dioxide
HyperSep Trypsin
HyperSep Zirconium Dioxide
Descrizione:
1 to 10 μL Volume
10 to 200 μL Volume
missing translation for 'unitSize'
pezzi 96
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Assorbente Descrizione unitSize
10364513 BioBasic C18 1 to 10 μL Volume pezzi 96
10488684 BioBasic C8 1 to 10 μL Volume pezzi 96
10662094 BioBasic C4 1 to 10 μL Volume pezzi 96
10314043 HyperSep Hypercarb 1 to 10 μL Volume pezzi 96
10522075 HyperSep Hypercarb and C18 (Mix Mode) 1 to 10 μL Volume pezzi 96
10304673 HyperSep HILIC 1 to 10 μL Volume pezzi 96
10611016 HyperSep Trypsin 1 to 10 μL Volume pezzi 96
10610046 HyperSep Poros WAX 1 to 10 μL Volume pezzi 96
10620046 HyperSep Poros SAX 1 to 10 μL Volume pezzi 96
10534085 HyperSep Poros SCX 1 to 10 μL Volume pezzi 96
10522265 HyperSep Titanium Dioxide 1 to 10 μL Volume pezzi 96
10488874 BioBasic C8 10 to 200 μL Volume pezzi 96
10706334 BioBasic C4 10 to 200 μL Volume pezzi 96
10065694 HyperSep Zirconium Dioxide 1 to 10 μL Volume pezzi 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10364513 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 60109401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Este artículo no se puede devolver. Vea la política de devoluciones
Get faster sample preparation with minimal sample loss with these SPE micropipette tips and achieve separation in volumes as low as 100nL.
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Features:
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
  • [bullet] Filter-free design to eliminate dead volume
  • [bullet] No contamination from the supporting matrix
  • [bullet] Available in many popular chemistries
Applications include:
  • [bullet] Mass spectrometry
  • [bullet] Desalting
  • [bullet] Protein purification
  • [bullet] MALDI
  • [bullet] Electrophoresis
  • [bullet] HPCE
  • [bullet] HPLC
  • [bullet] CEC
TRUSTED_SUSTAINABILITY
Especificaciones
Descrizione 1 to 10 μL Volume
Da utilizzare con (applicazione) Microscale Solid Phase Extraction
Da utilizzare con (apparecchiatura) Micropipettes
Dimensione particelle 40 to 60 μm
Product Line HyperSep
Assorbente BioBasic C18
Tipo SpinTip
Volume (sistema metrico) 1 to 10 μL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.