Learn More
uvex Hypervision CA Foam Clear AF GY/GY/BU
Codice prodotto. 31751174 Sfoglia Tutto Uvex Prodotti
Cambia vista
Click to view available options
Lens Material:
Polycarbonate
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Lens Material Dimensione della confezione
31751174 Polycarbonate Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 31751174 Fornitore uvex N. del fornitore 9321615
  • ONLINE EXCLUSIVE PRICE
    16.95€
    16.51€ / pezzi 1
    Risparmia 0.44€
    2% Sconto
Spedizione stimata: 24-11-2026
Goggles uvex hypervision Clear 9321615
  • The uvex hypervision features a comfortable, ergonomic design with a translucent frame that provides an almost unrestricted field of vision for enhanced workplace awareness.
  • A soft, flexible frame construction ensures high wearer comfort, even during extended periods of use.
  • Integrated side recesses allow sufficient space for personal prescription spectacles, enabling the uvex hypervision to be worn comfortably as reliable overspecs.
  • An adjustable headband combined with indirect ventilation completes the design, delivering a secure fit, improved airflow, and dependable eye protection for everyday professional applications.
TRUSTED_SUSTAINABILITY
Specifica
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Anti-fog Goggles
Materiale Polycarbonate
Monouso Reusable
Lens Material Polycarbonate
Materiale del telaio Polycarbonate
UV Protection UV380
Frame Type Standard