Descrizione
- The uvex hypervision features a comfortable, ergonomic design with a translucent frame that provides an almost unrestricted field of vision for enhanced workplace awareness.
- A soft, flexible frame construction ensures high wearer comfort, even during extended periods of use.
- Integrated side recesses allow sufficient space for personal prescription spectacles, enabling the uvex hypervision to be worn comfortably as reliable overspecs.
- An adjustable headband combined with indirect ventilation completes the design, delivering a secure fit, improved airflow, and dependable eye protection for everyday professional applications.
Specifica
Specifica
| Lens Coating | Scratch-resistant, Impact-resistant, Anti-fog |
| Lens Tint | None |
| Product Type | Goggles |
| Materiale | Polycarbonate |
| Monouso | Reusable |
| Lens Material | Polycarbonate |
| Materiale del telaio | Polycarbonate |
| UV Protection | UV400 |
| Frame Type | Classic |