Learn More
DiscoverX Ibrutinib (PCI-32765)
Codice prodotto. 17362559
Encompass
Cambia vista
Click to view available options
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
17362559 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17362559 Fornitore DiscoverX N. del fornitore 921243
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Ibrutinib (PCI-32765) is an inhibitor of BTK, YES, and other SRC tyrosine kinases. Provided as 5mg of dry powder.
Ibrutinib (PCI-32765, Imbruvica) is an FDA approved drug that targets B-cell malignancies and is approved for treatment of mantle cell lymphoma, chronic lymphocytic leukemia, and Waldenstrom's macroglobulinemia. Ibrutinib is an orally-bioavailable compound that is a potent and selective inhibitor of Bruton's tyrosine kinase (BTK). Also inhibits YES and other SRC family tyrosine kinases. Provided as 5mg of dry powder.
Specifica
Quantità 5 mg