missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibuprofen Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 38861-78-8 |
| Flash Point | 54.4°C |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. |
| Linear Formula | C12H16 O |
| Synonym | 4′-Isobutylacetophenone; 1-(4-Isobutylphenyl)ethanone; 1-Acetyl-4-isobutylbenzene |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 176.25 |
| Grade | Certified Reference Material |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?