missing translation for 'onlineSavingsMsg'
Learn More

ICAM-1 Mouse anti-Human, Clone: 2F9A8, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60299-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897375

  • 325.38€ / 150µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Intercellular adhesion molecule-1 (ICAM-1, CD54) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for lymphocyte function-associated (LFA) antigens, a beta 2 leukocyte integrin. The normal function of human ICAM-1 is to provide adhesion between endothelial cells and leukocytes after injury or stress. ICAM-1 binds to leukocyte function-associated antigen (LFA-1) or macrophage-1 antigen (Mac-1). It is found on leukocytes, fibroblasts, epithelial cells and endothelial cells and its expression is regulated by inflammatory cytokines. ICAM-1 has a tissue distribution similar to that of the major histocompatibility complex class II antigens and is likely to play a role in inflammatory responses.
Specifica

Specifica

ICAM-1
Monoclonal
0.7 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05362
ICAM1
ICAM-1 Fusion Protein Ag8309
150 μL
Primary
Human
Antibody
IgG2b
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
2F9A8
Unconjugated
ICAM1
BB2, CD54, ICAM 1, ICAM1, ICAM-1, P3.58
Mouse
Protein A
RUO
3383
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali