missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-3/CD50 Antibody (ICAM3/1019), Alexa Fluor™ 700, Novus Biologicals™
Sfoglia Tutto Bio Techne ProdottiDescrizione
ICAM-3/CD50 Monoclonal antibody specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifica
Specifica
| Antigene | ICAM-3/CD50 |
| Applicazioni | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classificazione | Monoclonal |
| Clone | ICAM3/1019 |
| Coniugato | Alexa Fluor 700 |
| Formulazione | 50 mM Sodium Borate |
| Alias gene | CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 |
| Specie ospite | Mouse |
| Immunogeno | Recombinant human ICAM-3/CD50 protein (Uniprot: P32942) |
| Metodo di purificazione | Protein A or G purified |
| Vedi altri risultati |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?