missing translation for 'onlineSavingsMsg'
Learn More

Icomethasone 21-Acetate, TRC

Product Code. 30134761
Change view
Click to view available options
Quantità:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30134761 50 mg 50mg
30134851 100 mg 100mg
18021440 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134761 Supplier Toronto Research Chemicals Supplier No. TRCI16410550MG
 Articolo soggetto a restrizioni
Spedizione stimata: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24916-91-4
Formula molecolare C24 H31 Cl O6
Molecular Weight (g/mol) 450.95
Sinonimo Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-9-chloro-11,17-dihydroxy-16-methyl-, (11β,16α)- (9CI), Pregna-1,4-diene-3,20-dione, 9-chloro-11β,17,21-trihydroxy-16α-methyl-, 21-acetate (7CI,8CI), Icometasone-21-acetate, Icometasone 21-Acetate, Mometasone Furoate Imp. O (EP)
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C

Specifications

Materiale o nome chimico Icomethasone 21-Acetate
Quantità 50 mg
InChI Formula InChI=1S/C24H31ClO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,19+,21+,22+,23+,24+/m1/s1
Peso formulazione 450.1809
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.