Learn More
MP Biomedicals™ IgG Affinity Gel
Product Code. 11424295 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424295 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11424295 Supplier MP Biomedicals™ Supplier No. 0855961
  • ONLINE EXCLUSIVE PRICE
    306.00€
    290.85€ / 2mL
    Risparmia 15.15€
    4% Sconto
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
IgG Affinity Gel is prepared by reaction of purified IgG to activated Sepharose™ 4B gel.
  • Product is rabbit IgG (whole molecule) covalently coupled to Sepharose-4B
  • Packed in a polystyrene chromatography column
Specifications
Binding Capacity 2.3 mg Antibody/mL
Quantity 2 mL
Content And Storage 2°C to 8°C
For Use With (Application) For affinity purification of antibodies to Rabbit IgG or to remove these antibodies from an antiserum preparation
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.