Learn More
Rabbit anti-Sheep IgG (H+L) Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marca: Invitrogen SA510311
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
This antibody will react with sheep IgG and with light chains common with other sheep immunoglobulins as determined by ELISA and IEP techniques. This antibody may cross-react with IgG from other species.
Sheep IgG (H+L) Polyclonal antibody specifically detects Sheep IgG (H+L) in Ovine samples. It is validated for ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot.Specifica
Sheep IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Ovine | |
Liquid |
ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Ovine IgG h+l | |
1 mg | |
Secondary | |
4° C | |
IgG |