missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-21 Polyclonal Antibody

Codice prodotto. 15567452
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15567452 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15567452 Fornitore Invitrogen™ N. del fornitore PA546962

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Goat Polyclonal Antibody

In sandwich ELISAs, less than 0.5% cross-reactivity with recombinant mouse (rm) IL-2, rmIL-4, rmIL-15, and recombinant human IL-21 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.30 EU per 1 μg of the antibody by the LAL method.

IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.
TRUSTED_SUSTAINABILITY

Specifica

Antigene IL-21
Applicazioni ELISA, Neutralization, Western Blot, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 0.2 mg/mL
Coniugato Unconjugated
Formulazione PBS with 5% trehalose and No Preservative
Gene IL21
N. accesso geni Q9ES17
Alias gene CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11
Simboli geni IL21
Specie ospite Goat
Immunogeno E. coli-derived recombinant mouse IL-21 Pro25-Ser146.
Metodo di purificazione Antigen affinity chromatography
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 60505
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Forma Lyophilized
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.