Learn More
IL1 beta Rabbit anti-Human, Polyclonal, Proteintech
Codice prodotto. 16886544 Sfoglia Tutto Proteintech Prodotti
Cambia vista
Click to view available options
Quantità:
20 μL
150 μL
Dimensione della confezione:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16886544 150 μL 150µL
16896544 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 16886544 Fornitore Proteintech N. del fornitore 168061AP150UL
Rabbit Polyclonal Antibody
Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
Specifica
Antigene IL1 beta
Applicazioni Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classificazione Polyclonal
Concentrazione 0.19 mg/mL
Coniugato Unconjugated
Formulazione PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL1B
N. accesso geni P01584
Alias gene IL1 beta, Catabolin, IL 1, IL 1 beta, IL1 BETA, IL1B, IL1beta, IL1F2, Interleukin 1 beta, interleukin 1, beta, Pro IL-1 beta
Simboli geni IL1B
Specie ospite Rabbit
Immunogeno IL1 beta Fusion Protein Ag10295
Metodo di purificazione Antigen Affinity Chromatography
Quantità 150 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 3553
Target Species Human
Content And Storage -20°C
Product Type Antibody
Forma Liquid
Isotype IgG