Learn More
Thermo Scientific™ Imidazole, 0.5M buffer soln., pH 6.0
Codice prodotto. 15414439
Cambia vista
Click to view available options
Quantità:
100 mL
250 mL
Dimensione della confezione:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15414439 100 mL 100mL
15424439 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15414439 Fornitore Thermo Scientific™ N. del fornitore J61956.AE
Richiesta di formato massivo o personalizzato
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifica
CAS 288-32-4
Colore Colorless
Concentrazione 0.5M buffer soln.
pH 6.0
Forma fisica Liquid
Quantità 100 mL
Formula molecolare C3H4N2
Numero MDL MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.079