Learn More
Thermo Scientific™ Imject™ Maleimide-Activated Blue Carrier™ Protein
Codice prodotto. 11899933 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
2 mg
10 mg
10 x 10 mg
Dimensione della confezione:
10mg
2mg
pezzi 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11899933 10 x 10 mg pezzi 10
11821184 2 mg 2mg
11810094 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Normativo: Questo prodotto contiene una sostanza elencata nell'allegato XVII del regolamento REACH (CE) n. 1907/2006. Tutti i clienti devono compilare una dichiarazione prima della consegna.
Codice prodotto. 11899933 Fornitore Thermo Scientific™ N. del fornitore 77662
 Articolo soggetto a restrizioni
Spedizione stimata: 07-12-2026
Normativo: Questo prodotto contiene una sostanza elencata nell'allegato XVII del regolamento REACH (CE) n. 1907/2006. Tutti i clienti devono compilare una dichiarazione prima della consegna.
Make hapten-carrier immunogens comprising cysteine-containing peptides and other thiolated antigens with sulfhydryl-reactive immunogenic hemocyanin (CCH).
Thermo Scientific™ Maleimide-Activated Blue Carrier Protein is sulfhydryl-reactive immunogenic hemocyanin (CCH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Blue Carrier Protein is a large homocyanin that is highly immunogenic like keyhole limpet hemocyanin (KLH) yet is more soluble. The carrier is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Numerous antigen molecules can be conjugated to each Blue Carrier Protein molecule to make them highly immunogenic and increase the likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest.
Highlights:
  • Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of carrier, ensuring consistently high levels of conjugatio
  • Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
  • Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
  • High-quality carrier protein – Blue Carrier Protein is ultrapure Concholepas concholepas hemocyanin (CCH) prepared from molluscan hemolymph
  • Highly immunogenic and soluble – CCH has a high molecular mass that elicits a stronger immune response than BSA or ovalbumin
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Upon receipt store at -20°C. Product is shipped on ice.
Quantità 10 x 10 mg
Product Line Blue Carrier, Imject
Product Type Imject Maleimide-Activated Blue Carrier Protein