Learn More
Fisherbrand™ Immersion Basket with Handle
Product Code. 11758728 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Ultrasonic Bath, Elmasonic X-tra ST 1400H
Ultrasonic Bath, Elmasonic Xtra Basic 1600 160 L
Ultrasonic Bath, Elmasonic Xtra Basic 2500 250 L
Ultrasonic Bath, Elmasonic Xtra Basic 300 30 L
Ultrasonic Bath, Elmasonic Xtra Basic 550 55L
Ultrasonic Bath, Elmasonic Xtra Basic 800 80 L
Ultrasonic Cleaner, Elmasonic S450 H 45 L
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11758728 Ultrasonic Bath, Elmasonic Xtra Basic 300 30 L Each
11786198 Ultrasonic Cleaner, Elmasonic S450 H 45 L Each
11778728 Ultrasonic Bath, Elmasonic Xtra Basic 550 55L Each
11788728 Ultrasonic Bath, Elmasonic Xtra Basic 800 80 L Each
11708738 Ultrasonic Bath, Elmasonic X-tra ST 1400H Each
11718738 Ultrasonic Bath, Elmasonic Xtra Basic 1600 160 L Each
11758738 Ultrasonic Bath, Elmasonic Xtra Basic 2500 250 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11758728
  • ONLINE EXCLUSIVE PRICE
    261.00€
    245.00€ / pezzi 1
    Risparmia 16.00€
    6% Sconto
View all promotions
Spedizione stimata: 04-01-2027
Add to basket
Add to basket
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
  • Mesh bottom
  • One handle on each end
  • Functional and modern design
  • Available for a variety of capacities
Specifications
Product Type Basket
For Use With (Equipment) Ultrasonic Bath, Elmasonic Xtra Basic 300 30 L
Material Stainless Steel