missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Immuno Standard Modules, EB, black
Click to view available options
Dimensione della confezione:
pezzi 50
Descrizione
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Features:
- Available in eight-, 12- or 16-well strips
- Framed or unframed for choice of optimal assay
- Flat-bottom or C-shaped wells
- Choice of three surface types for optimal binding
- White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
- Black format to reduce back scatter light which may be encountered in epifluorescence
- Transparent or clear format for use in Time Resolved Fluorescence applications only
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
- Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.
Specifications
Specifications
| Proprietà di legame | Hydrophilic |
| Certificato | No |
| Formato | Strip Plate |
| Metodo di rilevamento | Fluorescence |
| Matrice | 8 x 12 |
| Imballaggio | 50 Boxes |
| Colore | Black |
| Product Type | Microplate Strip |
| Materiale | Polystyrene |
| No. of Wells | 96 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction