missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX InCELL Hunter™ A549 CSF1R Kinase Binding Cell Line

Codice prodotto. 17384789
Encompass
Cambia vista
Click to view available options
Quantità:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
17384789 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17384789 Fornitore DiscoverX N. del fornitore 960047C15

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The InCELL Hunter™ A549 CSF1R Kinase Binding Cell Line measures target engagement via compound-induced stabilization of CSF1R target protein. This cell line ships as two vials of cryopreserved cells.

The InCELL Hunter™ A549 CSF1R Kinase Binding Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with kinases. This cell line enables assessment of compound-engagement-based stabilization of CSF1R via detection of CSF1R levels. This product uses EFC technology. The cell line included overexpresses CSF1R target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is denatured by the heat treatment and no EFC signal is produced. When compound engages and heat-stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifica

Assay Entry Target Engagement
Linea cellulare A549
Da utilizzare con (applicazione) Cell Lines
Quantità 2 Vials
Target Entry CSF1R
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.