Learn More
Aqualytic™ Incubator, thermostatic control +2°C to +40°C, with LED display, 255L capacity
Marca: Aqualytic™ 438230
Dettagli aggiuntivi : Peso : 85.00000kg
Descrizione
- Temperature range +2°C to +40°C, adjustable in steps of 0.1°C
- Low power consumption
- Illuminated LED Display
- Power sockets inside incubator
- Delivered with bottom grid