missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indiko™ System Albumin to CRP Specific Protein Reagents

Codice prodotto. 17773627 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Analiti rilevabili:
CRP
Formato:
Antiserum Kit
Calibration Set
Quantità:
1 x 3 mL
5 x 1 mL
Dimensione della confezione:
3mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Analiti rilevabili Formato Quantità unitSize
17773627 CRP Calibration Set 5 x 1 mL 5mL
17783637 CRP Antiserum Kit 1 x 3 mL 3mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17773627 Fornitore Thermo Scientific™ N. del fornitore 981674
 Articolo soggetto a restrizioni
Spedizione stimata: 12-08-2026
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Increase testing efficiency with our ready-to-use, barcoded Thermo Scientific™ Indiko™ System Albumin to CRP Specific Protein Reagents.

Receive best-in-class methodologies and standardized performance with Thermo Scientific™ Indiko™ System Specific Protein Reagents, which include a specific protein test menu of over 20 immunoturbidimetric methods. The assays are always ready to go thanks to full validation and compliance as well as barcoding.

All Indiko methods are based on carefully designed protocols for high reagent stability to support less calibration, optimized QC, and fewer repeats. Calibrators and controls are available in separate kits.

Highlights:
  • High-quality reagents are barcoded for automatic identification
  • Most reagents are delivered in ready-to-use liquid form
  • Fully validated applications in accordance with the applicable EU regulatory requirements for IVD products permit holistic quality assurance in your lab

Ordering Information: Not available in the U.S.A.

Specific Protein Applications for Indiko™ Analyzers:

  • Albumin / Microalbuminuria
  • Antistreptolysin O
  • Alpha-1-antitrypsin
  • Alpha-1-glycoprotein acid
  • Apolipoprotein A1 & B
  • Complement C3 and C4
  • C-reactive protein
  • CRP High Sensitivity
  • D-Dimer
  • Ferritin
  • HbA1c
  • Haptoglobin
  • IgA, G & M
  • Lipoprotein(a)
  • Rheumatoid factors
  • Soluble transferrin receptor
  • Transferrin
TRUSTED_SUSTAINABILITY

Specifica

Descrizione CRP Calibration Set; Kit size: 5 x 1mL
Analiti rilevabili CRP
Formato Calibration Set
Tipo di campione Serum
Tipo Reagent
Quantità 5 x 1 mL
Forma Liquid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.