Learn More
Thermo Scientific™ Indiko™ System Calibrators and Controls
Codice prodotto. 10156924 Sfoglia Tutto Thermo Scientific Prodotti
Encompass
Cambia vista
Click to view available options
Analiti rilevabili:
sCal Calibrator
Dimensione della confezione:
pezzi 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Analiti rilevabili Dimensione della confezione
10156924 sCal Calibrator pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10156924 Fornitore Thermo Scientific™ N. del fornitore 981831
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
missing translation for 'validMainframeMsg'
Enable accurate test results with Thermo Scientific™ Indiko™ System Calibrators and Controls.
Enable accurate results with Thermo Scientific™ Indiko™ System Calibrators and Controls. Indiko analyzers are fully automated for routine and specialty testing. Ready-to-use barcoded reagents are designed to support efficient operations and reduce the risk of errors.
Highlights:
  • Based on serum or urine of human or bovine origin
  • Provided in a lyophilized form
Ordering information:
  • Nortrol, Abtrol and sCal are available in the U.S.A.
  • All other calibrators and controls are not available in the U.S.A.
Specifica
Certificazione/Conformità Compliant with the IVDR; CE marked
Analiti rilevabili sCal Calibrator
Tipo di campione Serum, Plasma, Urine
Tipo Calibrator
Quantità 10 x 3 mL