missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Enhance your laboratory's performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.
Indiko™ applications utilize carefully selected methods according to IFCC and other widely used recommendations. Clinical chemistry methods are provided with system specified multi-calibrators and -controls with traceability to international reference materials.
Ordering Information: Not available in the U.S.A., except Glucose Hexokinase.
Clinical Chemistry Applications for Indiko™ Analyzers:
Ordering Information: Not available in the U.S.A., except Glucose Hexokinase.
Clinical Chemistry Applications for Indiko™ Analyzers:
- Acid Phosphatase
- Albumin
- ALP
- ALT
- Amylase
- AST
- Bilirubin direct and total
- Calcium
- Cholesterol
- HDL and LDL Cholesterol
- Cholinesterase
- CO2
- Creatine Kinase
- CK-MB
- Creatinine
- Gamma-GT
- Glucose
- alpha-HBDH
- Iron
- LDH
- Lipase
- Magnesium
- Phosphorus
- Total Protein
- Triglycerides
- U/CSF Protein
- Urea
- Uric Acid
Specifications
Specifications
| Certificazione/Conformità | CE Marked |
| Descrizione | Cholesterol; Kit Size: 12 x 20mL |
| Marchio CE | Yes |
| Analiti rilevabili | Cholesterol |
| Tipo di campione | Serum, Urine |
| Tipo | Reagent |
| Quantità | 12 x 20 mL |
| Forma | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?