missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents, Alkaline Phosphatase to Creatinine

Product Code. 11985152 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantità:
15 x 5 mL
20 x 4 mL
4 x 30 mL
7 x 50 mL
8 x 20 mL
Analiti rilevabili:
ALT/GPT
AST/GOT
Alkaline Phosphatase
Creatinine (Enzymatic)
Direct Bilirubin
Unit Size:
120mL
160mL
75mL
80mL
Pezzo
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Analiti rilevabili unitSize
11985152 7 x 50 mL AST/GOT Pezzo
17783627 8 x 20 mL ALT/GPT 160mL
17793627 8 x 20 mL AST/GOT 160mL
17753637 15 x 5 mL Alkaline Phosphatase 75mL
17773637 4 x 30 mL Creatinine (Enzymatic) 120mL
17763637 20 x 4 mL Direct Bilirubin 80mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11985152 Supplier Thermo Scientific™ Supplier No. 981363
  • ONLINE EXCLUSIVE PRICE
    250.00€
    160.84€ / pezzi 1
    Risparmia 89.16€
    35% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Standardize performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents, which offer best-in-class methodologies.

Enhance your laboratory's performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.

Features:
  • Indiko™ and Konelab™ applications utilize carefully selected methods according to IFCC and other widely used recommendations
  • Highly developed Indiko and Konelab analyzers are integrated systems for routine clinical chemistry, ISEs, specific proteins, TDMs and DATs
  • The CE marked kits are compliant with the IVDD
  • Clinical Chemistry methods are provided with system specified multi-calibrators and -controls with traceability to international reference materials
Ordering Information: Not available in the U.S.A., except Glucose Hexokinase.

Clinical Chemistry Applications for Indiko and Konelab Analyzers:
  • Acid Phosphatase
  • Albumin
  • ALP
  • ALT
  • Amylase
  • AST
  • Bilirubin direct and total
  • Calcium
  • Cholesterol
  • HDL and LDL Cholesterol
  • Cholinesterase
  • CO2
  • Creatine Kinase
  • CK-MB
  • Creatinine
  • Gamma-GT
  • Glucose
  • alpha-HBDH
  • Iron
  • LDH
  • Lipase
  • Magnesium
  • Phosphorus
  • Total Protein
  • Triglycerides
  • U/CSF Protein
  • Urea
  • Uric Acid
TRUSTED_SUSTAINABILITY

Specifications

Analiti rilevabili AST/GOT
Tipo di campione Serum
Tipo Reagent
Quantità 7 x 50 mL
Forma Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.