missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents, Alkaline Phosphatase to Creatinine

Codice prodotto. 17793627 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
15 x 5 mL
20 x 4 mL
4 x 25 mL
4 x 30 mL
7 x 50 mL
8 x 20 mL
Analiti rilevabili:
ALT/GPT
AST/GOT
Alkaline Phosphatase
Amylase
Creatinine (Enzymatic)
Direct Bilirubin
Dimensione della confezione:
120mL
160mL
75mL
80mL
Pezzo
pezzi 4
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Analiti rilevabili Dimensione della confezione
17793627 8 x 20 mL AST/GOT 160mL
11985152 7 x 50 mL AST/GOT Pezzo
17646315 4 x 25 mL Amylase pezzi 4
17783627 8 x 20 mL ALT/GPT 160mL
17753637 15 x 5 mL Alkaline Phosphatase 75mL
17773637 4 x 30 mL Creatinine (Enzymatic) 120mL
17763637 20 x 4 mL Direct Bilirubin 80mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17793627 Fornitore Thermo Scientific™ N. del fornitore 981771
  • ONLINE EXCLUSIVE PRICE
    99.20€
    91.50€ / 160mL
    Risparmia 7.70€
    7% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 30-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Standardize performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents, which offer best-in-class methodologies.

Enhance your laboratory's performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.

  • Indiko™ applications utilize carefully selected methods according to IFCC and other widely used recommendations
  • Highly developed Indiko analyzers are integrated systems for routine clinical chemistry and specialty testing
  • Fully validated applications in accordance with the applicable EU regulatory requirements for IVD products permit holistic quality assurance in your lab
  • Clinical Chemistry methods are provided with system specified multi-calibrators and -controls with traceability to international reference materials
Ordering Information:

Not available in the U.S.A., except Glucose Hexokinase.

Clinical Chemistry Applications for Indiko Analyzers:
  • Albumin
  • ALP
  • ALT
  • Ammonia
  • Amylase
  • AST
  • Bile acids TBA
  • Bilirubin direct and total
  • Calcium
  • Copper
  • Cholesterol
  • HDL and LDL Cholesterol
  • Cholinesterase
  • Bicarbonate
  • Creatine Kinase
  • CK-MB
  • Creatinine
  • Gamma-GT
  • Glucose
  • G6P-DH
  • Iron
  • Lactate
  • LDH
  • Lipase
  • Magnesium
  • Phosphorus
  • Total Protein
  • Triglycerides
  • U/CSF Protein
  • Urea
  • Uric Acid
  • Zinc
TRUSTED_SUSTAINABILITY

Specifica

Analiti rilevabili AST/GOT
Tipo di campione Serum
Tipo Reagent
Quantità 8 x 20 mL
Forma Liquid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.