Learn More
Thermo Scientific™ Indiko™ and Konelab™ System Glucose Reagents
Codice prodotto. 17703637 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
12 x 20 mL
Dimensione della confezione:
240mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
17703637 12 x 20 mL 240mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17703637 Fornitore Thermo Scientific™ N. del fornitore 981780
  • ONLINE EXCLUSIVE PRICE
    174.00€
    48.95€ / 240mL
    Risparmia 125.05€
    71% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 26-11-2026
Fully standardize performance with Thermo Scientific™ Indiko™ and Konelab™ System Glucose Reagents, which offer best-in-class methodologies.
Enhance your laboratory's performance with Thermo Scientific™ Indiko™ and Konelab™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.
Indiko™ and Konelab™ applications utilize carefully selected methods according to IFCC and other widely used recommendations. Clinical Chemistry methods are provided with system specified multi-calibrators and controls with traceability to international reference materials.
Ordering Information: Not available in the U.S.A., except Glucose Hexokinase.
The Clinical Chemistry Applications for Indiko and Konelab Analyzers cover:
  • Acid Phosphatase
  • Albumin
  • ALP
  • ALT
  • Amylase
  • AST
  • Bilirubin direct and total
  • Calcium
  • Cholesterol
  • HDL and LDL Cholesterol
  • Cholinesterase
  • CO2
  • Creatine Kinase
  • CK-MB
  • Creatinine
  • Gamma-GT
  • Glucose
  • alpha-HBDH
  • Iron
  • LDH
  • Lipase
  • Magnesium
  • Phosphorus
  • Total Protein
  • Triglycerides
  • U/CSF Protein
  • Urea
  • Uric Acid
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Glucose; Kit Size: 12 x 20mL
Analiti rilevabili Glucose
Tipo di campione Plasma, Serum
Tipo Reagent
Quantità 12 x 20 mL
Forma Liquid