missing translation for 'onlineSavingsMsg'
Learn More

Indoleacetic Acid-13C6, TRC

Product Code. 30390819
Change view
Click to view available options
Quantità:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
30390819 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390819 Supplier Toronto Research Chemicals Supplier No. TRCI5773421MG
 Articolo soggetto a restrizioni
Spedizione stimata: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 1H-Indole-3-acetic Acid 13C6 (indole-3a,4,5,6,7,7a-13C6)
CAS 100849-36-3
Formula molecolare 13C6 C4 H9 N O2
Quantità 1 mg
Sinonimo 1H-Indole-3a,4,5,6,7,7a-13C6-3-acetic acid, 1H-Indole-3-acetic acid 13C6 (indole-3a,4,5,6,7,7a-13C6), 2-([3a,4,5,6,7,7a-13C6]-1H-indol-3-yl)acetic acid, 2-(3-Indolyl-3a,4,5,6,7,7a-13C6)acetic acid, 3-Indolylacetic acid-13C6
InChI Formula InChI=1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)/i1+1,2+1,3+1,4+1,8+1,9+1
SMILES OC(=O)Cc1c[nH][13c]2[13cH][13cH][13cH][13cH][13c]12
IUPAC Name 2-(1H-indol-3-yl)acetic acid
Molecular Weight (g/mol) 181.14
Peso formulazione 181.083
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.