missing translation for 'onlineSavingsMsg'
Learn More

Indoleamine 2,3-dioxygenase/IDO Antibody (9E5), Novus Biologicals™

Codice prodotto. p-200022509 Shop All Bio Techne Products
Change view
Click to view available options
Quantità:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30034828 50 μL 50µL
30033983 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30034828 Supplier Novus Biologicals Supplier No. NBP32621050ul

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Indoleamine 2,3-dioxygenase/IDO Monoclonal antibody specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigene Indoleamine 2,3-dioxygenase/IDO
Applicazioni ELISA, Immunohistochemistry
Classificazione Monoclonal
Clone 9E5
Coniugato Unconjugated
Diluizione ELISA, Immunohistochemistry 1:50-1:200
Formulazione PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias gene EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
Specie ospite Rabbit
Immunogeno A synthesized peptide derived from Human Indoleamine 2,3-dioxygenase/IDO [UniProt P14902]
Metodo di purificazione Affinity purified
Quantità 50 μL
Status giuridico RUO
Disciplina di ricerca Cancer, Myeloid derived Suppressor Cell
Primario o secondario Primary
ID gene (immissione) 3620
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.