missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Influenza A[A/swine/Jiangsu/J004/2018(H1N1)] Neuraminidase (NA) Protein, His Tag

dietro preventivo

Specifica

Numero di accesso QEM24763.1
Peso molecolare 56.8 kDa
Sorgente Influenza A[A/swine/Jiangsu/J004/2018(H1N1)] NA, His Tag (NEE-V5243) is expressed from human 293 cells (HEK293). It contains AA Ile 30 - Lys 469 (Accession # QEM24763.1).
Immunogeno Ile 30 - Lys 469
Requisiti di stoccaggio -20°C
Visualizza altre specifiche

Descrizione

Descrizione

Influenza, commonly known as “the flu”, is an infectious disease of birds and mammals caused by RNA viruses of the family Orthomyxoviridae, the influenza viruses. The virus is divided into three main types (Influenzavirus A, Influenzavirus B, and Influenzavirus C), which are distinguished by differences in two major internal proteins (hemagglutinin (HA) and neuraminidase (NA), which are the most important targets for the immune system. The type A viruses are the most virulent human pathogens among the three influenza types and cause the most severe disease. The serotypes that have been confirmed in humans, ordered by the number of known human pandemic deaths, are: H1N1, H2N2, H3N2, H5N1, H7N7, H1N2, H9N2, H7N2, H7N3, H10N7, H7N9. H7N9 is a serotype of the species Influenzavirus A (avian influenza virus or bird flu virus). H7 normally circulates amongst avian populations with some variants known to occasionally infect humans. An H7N9 virus was first reported to have infected humans in 2013 in China.

This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 56.8 kDa.
Specifica

Specifica

QEM24763.1
Influenza A[A/swine/Jiangsu/J004/2018(H1N1)] NA, His Tag (NEE-V5243) is expressed from human 293 cells (HEK293). It contains AA Ile 30 - Lys 469 (Accession # QEM24763.1).
-20°C
Neuraminidase (NA)
Influenza
Less than 1.0 EU / µg by the LAL method.
Powder
56.8 kDa
Ile 30 - Lys 469
NA,Neuraminidase
Unconjugated
His Tag
HEK293
Viral Proteins
Videos
SDS
Documenti

Documenti

Product Certifications