Learn More
ACROBiosystems Influenza A[A/swine/Jiangsu/J004/2018(H1N1)] Neuraminidase (NA) Protein, His Tag
Marca:  ACROBiosystems NEE-V5243-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17106541
  • dietro preventivo / 100µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
Influenza, commonly known as “the flu”, is an infectious disease of birds and mammals caused by RNA viruses of the family Orthomyxoviridae, the influenza viruses. The virus is divided into three main types (Influenzavirus A, Influenzavirus B, and Influenzavirus C), which are distinguished by differences in two major internal proteins (hemagglutinin (HA) and neuraminidase (NA), which are the most important targets for the immune system. The type A viruses are the most virulent human pathogens among the three influenza types and cause the most severe disease. The serotypes that have been confirmed in humans, ordered by the number of known human pandemic deaths, are: H1N1, H2N2, H3N2, H5N1, H7N7, H1N2, H9N2, H7N2, H7N3, H10N7, H7N9. H7N9 is a serotype of the species Influenzavirus A (avian influenza virus or bird flu virus). H7 normally circulates amongst avian populations with some variants known to occasionally infect humans. An H7N9 virus was first reported to have infected humans in 2013 in China.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 56.8 kDa.
Specifica
Specifica
QEM24763.1
56.8 kDa
Influenza A[A/swine/Jiangsu/J004/2018(H1N1)] NA, His Tag (NEE-V5243) is expressed from human 293 cells (HEK293). It contains AA Ile 30 - Lys 469 (Accession # QEM24763.1).
-20°C
Neuraminidase (NA)
His Tag
HEK293
Influenza
Powder
100 µg
Ile 30 - Lys 469
NA,Neuraminidase
Unconjugated
Less than 1.0 EU / µg by the LAL method.
Viral Proteins