Learn More
Mettler Toledo™ InLab™ OptiOx Pro-DES™ DO BOD Adapter
GREENER_CHOICE
Codice prodotto. 31454068 Sfoglia Tutto Mettler Toledo Prodotti
Cambia vista
Click to view available options
Tipo:
BOD Adapter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo
31454068 BOD Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 31454068 Fornitore Mettler Toledo™ N. del fornitore 30786584
Enables easy installation of the sensor for use in common BOD applications.
  • Use the BOD adapter to install the InLab OptiOx Pro-DES sensor on all common BOD applications.
TRUSTED_SUSTAINABILITY
Specifica
Tipo BOD Adapter