Descrizione
- Optical sensing cap compatible with InLab OptiOx Pro-DES sensor, which can be purchased separately for replacement; it has a lifespan of 18 months, ensuring long-term usability and cost-effective maintenance
Specifica
Specifica
| Tipo | RDO Fast Cap Replacement Kit |