Learn More
Novus Biologicals™ InnovaCoat (R) GOLD 80nm Midi Antibody Labeling Kit
Codice prodotto. 18075085 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
1 Test
Dimensione della confezione:
1 test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18075085 1 Test 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only
Codice prodotto. 18075085 Fornitore Novus Biologicals™ N. del fornitore 2310015
 Articolo soggetto a restrizioni
Spedizione stimata: 01-10-2026
Aggiungi al carrello
Aggiungi al carrello
For Research Use Only
Use simple pipetting operations to covalently attach antibodies, proteins and other biomolecules to ultra-stable InnovaCoat GOLD nanoparticles. Hands-on time is about two minutes and the conjugate is ready within fifteen.
Features
  • Proprietary surface coat reduces risk of sub-optimal antibody performance
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Antibody-GOLD conjugates in just 15 minutes
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Coating procedure not dependent on isoelectric point of antibody
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Store at 4°C. Do not freeze.
Quantità 1 Test
Product Type Antibody Labeling Kit