Learn More
Miele Professional™ Insert
Product Code. 15430565 Shop All Miele Products
Change view
Click to view available options
Holds:
46 Erlenmeyer flasks
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
15430565 46 Erlenmeyer flasks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15430565 Supplier Miele Professional™ Supplier No. 5892360
  • ONLINE EXCLUSIVE PRICE
    1630.00€
    1500.13€ / pezzi 1
    Risparmia 129.87€
    7% Sconto
Spedizione stimata: 01-02-2027
Mobile unit E 960/1 for the optimum loading of wide necked Erlenmeyer flasks
  • Equipped with 20 large and 26 small spring hooks
  • Suitable for Erlenmeyer flasks and measuring cylinders
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Product Type Insert/Module for Wide-necked Glassware
Color Stainless Steel
Holds 46 Erlenmeyer flasks
For Use With (Equipment) Large-capacity washer-disinfectors, laboratory
Depth (Metric) 522 mm
Height (Metric) 185 mm
Width (Metric) 357 mm