Learn More
Hanna Instruments™ Kit per test a reagente delle acque Instrument
Click to view available options
Range:
0 - 0.5 mg/l
0 to 500 mg/L Total
0.00 to 10.00 mg/L Free and Total
0.00 to 5.00 mg/L Total
0.000 to 0.500 mg/L Free
N. di test:
100
300
600
Da utilizzare con (applicazione):
Chlorine Ultra High Range
Free Chlorine Ultra Low Range
Free and Total Chlorine High Range
Total Chlorine
Total Chlorine (DPD3)
Total Chlorine Ultra Low Range
Dimensione della confezione:
Pezzo
pezzi 100
Descrizione
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
- Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifica
Specifica
| Da utilizzare con (applicazione) | Free Chlorine Ultra Low Range |
| Da utilizzare con (apparecchiatura) | Compatible Hanna benchtop and portable photometers |
| Include | 100 individual packets of HI95762-0 |
| N. di test | 100 |
| Quantità | 100/conf. |
| Range | 0 - 0.5 mg/l |
| Metodo test | Standard Method 4500-Cl G |
| Tipo | Reagente |