Learn More
Integrated Gas Module for Varioskan™ LUX
Control O2 and CO2 levels with this optional, integrated gas module for the Thermo Scientific™ Varioskan™ LUX multimode microplate reader.
Marca:  Thermo Scientific VLUPG006
Dettagli aggiuntivi : Peso : 1.20000kg
Codice prodotto. 15320787
  • 11148.00€ / Pezzo
Spedizione stimata
26-09-2018
Aggiungi al carrello
Specifica
Specifica
Integrated gas module upgrade kit (service upgradable)
0.1 to 15%
1 to 21%
Varioskan LUX
± 0.3% at 5% CO2
±0.3% at 1% O2
Videos
NOTE: Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.