Learn More
Integrin alpha 4/CD49d Antibody (CL11294) [FITC], Novus Biologicals™
Codice prodotto. 30294657 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30294657 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30294657 Fornitore Novus Biologicals N. del fornitore NBP307999F
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
Integrin alpha 4/CD49d Monoclonal antibody specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifica
Antigene Integrin alpha 4/CD49d
Applicazioni Western Blot
Classificazione Monoclonal
Clone CL11294
Coniugato FITC
Formulazione PBS
Alias gene 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Specie ospite Mouse
Immunogeno This antibody was developed using a synthetic peptide derived from P13612, with the exact immunogen sequence remaining proprietary.
Metodo di purificazione Protein A purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Cancer, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Microautophagy, Neuronal Cell Markers, Phospho Specific
Primario o secondario Primary
ID gene (immissione) 3676
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Forma Purified
Isotype IgG2b