missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18309-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
ITGB3 is involved in Glanzmann thrombasthenia (GT), a common inherited disease of platelet aggregation. It is characterized by mucocutaneous bleeding of mild-to-moderate severity. GT has been classified clinically into types I and II. In type I, platelets show absence of the glycoprotein IIb-IIIa complexes at their surface and lack fibrinogen and clot retraction capability. In type II, the platelets express the GPIIb-IIIa complex at reduced levels, have detectable amounts of fibrinogen, and have low or moderate clot retraction capability. The disease is caused by mutations affecting the gene represented in this entry.Specifica
Integrin beta-3 | |
Polyclonal | |
Unconjugated | |
ITGB3 | |
CD61, GP3A, GPIIIa, Integrin beta 3, Integrin beta-3, ITGB3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16416, 29302, 3690 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O54890, P05106 | |
ITGB3 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |