Learn More
Thermo Scientific™ (IOQ) Qualification Service
Codice prodotto. 30049054 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Product Type:
IQ/OQ Service
Dimensione della confezione:
1 unità
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type Dimensione della confezione
30049054 IQ/OQ Service 1 unità
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30049054 Fornitore Thermo Scientific™ N. del fornitore IOQPCKE89003711
 Articolo soggetto a restrizioni
Spedizione stimata: 24-11-2026
Qualification service for New Heratherm Chamber 750 L (including documentation).
  • IQ: Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements.
  • Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order.
  • Verifies equipment and any other accessories are assembled and installed.
  • Configuration information for each system component gets recorded. Installation of Equipment is NOT Included.
  • OQ: Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
  • Key tests: Door Ajar, Power Failure, Temperature Alerts
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (apparecchiatura) Thermo Scientific New Heratherm Chamber 750 L
Product Type IQ/OQ Service